Would you like to log in again?
Human Recombinant Aldo-Keto Reductase 1C4 (from E. coli)
Supplier: ProSci
92-002
75788-676EA 946.19 USD InStock
75788-676
Human Recombinant Aldo-Keto Reductase 1C4 (from E. coli)
Enzymes
Aldo-Keto Reductase 1C4/AKR1C4 is a member of the aldo/keto reductase family that consists of more than 40 known enzymes and proteins
This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: N-6 His tag
AKR1C4 has highly expressed in Liver. It can catalyzes the bioreduction of chlordecone, a toxic organochlorine pesticide, to chlordecone alcohol in liver. AKR1C4 catalyzes the transformation of the potent androgen dihydrotestosterone (DHT) into the less active form, 5- alpha-Androstan-3- alpha,17- beta -diol (3- alpha-diol). In addition, AKR1C4 also has some 20- alpha-Hydroxysteroid Dehydrogenase activity.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty