Would you like to log in again?
Human Recombinant Sulfotransferase 1A2 (from E. coli)
Supplier: ProSci
92-054
75790-702EA 744.04 USD InStock
75790-702
Human Recombinant Sulfotransferase 1A2 (from E. coli)
Enzymes
Sulfotransferase 1A2 (SULT1A2) is a member of the Sulfotransferase 1 family
This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: N-6 His tag
Sulfotransferase enzymes catalyze the sulfate conjugation of many hormones, neurotransmitters, drugs, and xenobiotic compounds. SULT1A2 is a cytoplasmic protein and exists as a homodimer. SULT1A2 mediates the metabolic activation of carcinogenic N-hydroxyarylamines to DNA binding products and might thus participate as a modulating factor of cancer risk.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty