Would you like to log in again?
Human Recombinant Deoxycytidine Kinase (from E. coli)
Supplier: ProSci
92-064
75790-722EA 744.04 USD InStock
75790-722
Human Recombinant Deoxycytidine Kinase (from E. coli)
Enzymes
Deoxycytidine Kinase (DCK) is a member of the DCK/DGK family
This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: N-6 His, T7 tag
DCK exists as a homodimer and is localized to the nucleus. DCK is required for the phosphorylation of the deoxyribonucleosides deoxycytidine (dC), deoxyguanosine (dG), and deoxyadenosine (dA). DCK has broad substrate specificity, and does not display selectivity based on the chirality of the substrate. In addition, DCK is also an essential enzyme for the phosphorylation of numerous nucleoside analogs widely employed as antiviral and chemotherapeutic agents. DCK is clinically important because of its relationship to drug resistance and sensitivity.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty