Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Human recombinant ribulose-phosphate 3-epimerase (from E. coli)
Supplier: ProSci
92-090
75790-774EA 700.27 USD InStock
75790-774
Human recombinant ribulose-phosphate 3-epimerase (from E. coli)
Enzymes
Ribulose-Phosphate 3-Epimerase (RPE) is a member of the Ribulose-Phosphate 3-Epimerase family
This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
RPE exists as a homodimer and catalyzes the reversible epimerization of D-ribulose 5-phosphate to D-xylulose 5-phosphate. RPE binds one divalent metal cation per subunit and contains tightly bound Fe2+ when produced in E. coli, but the physiological cofactor may be Co2+, Mn2+ or Zn2+. It has been shown that RPE participates in 3 metabolic pathways: pentose phosphate pathway, pentose and glucuronate interconversions, and carbon fixation.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty