Would you like to log in again?
Human Recombinant Bisphosphoglycerate Mutase (from E. coli)
Supplier: ProSci
92-092
75790-778EA 744.04 USD InStock
75790-778
Human Recombinant Bisphosphoglycerate Mutase (from E. coli)
Enzymes
Bisphosphoglycerate Mutase (BPGM) is a member of the Phosphoglycerate Mutase family and BPG-Dependent PGAM subfamily
This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
BPGM is a multifunctional enzyme. BPGM catalyzes 2,3-DPG synthesis via its synthetase activity, and 2,3-DPG degradation via its phosphatase activity. It also has phosphoglycerate phosphomutase activity. BPGM plays a major role in regulating hemoglobin oxygen affinity by controlling the levels of 2,3-bisphosphoglycerate (2,3-BPG). Deficiency of BPGM increases the affinity of cells for oxygen and result in hemolytic anemia.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty