Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Human Recombinant Uridine Phosphorylase 1 (from E. coli)
Supplier: ProSci
92-122
75790-838EA 700.27 USD InStock
75790-838
Human Recombinant Uridine Phosphorylase 1 (from E. coli)
Enzymes
Uridinephosphorylase 1 (UPP1) is a member of the family of pentosyltransferase
This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: N-6 His tag
UPP1 catalyses the reversible phosphorolysis of uridine to uracil. The expression levels and the enzymatic activity of UPP1 are higher in human solid tumors than in adjacent normal tissues. The high level of UPP1 expression in some tumors makes it a potential prognosticfactor for some cancers, such as oral squamous cell carcinoma. UPP1 is important for the homeostatic regulation of intracellular and plasma uridine concentratios. UPP1 plays an important role in the pyrimidine salvage pathway through its catalysis of the reversible phosphorolysis of uridine to uracil.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty