Would you like to log in again?
Human Recombinant Sulfotransferase 1B1 (from E. coli)
Supplier: ProSci
92-131
75790-856EA 946.47 USD InStock
75790-856
Human Recombinant Sulfotransferase 1B1 (from E. coli)
Enzymes
Sulfotransferase Family Cytosolic 1B Member 1 (SULT1B1) is a cytosolic enzyme that belongs to the Sulfotransferase 1 family
This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: N-6 His tag
Human SULT1B1 is a 296 amino acid protein that is highly expressed in the liver, peripheral blood leukocytes, colon, small intestine, and spleen. SULT1B1 utilizes 3'-phospho-5'-adenylyl sulfate (PAPS) as sulfonate donor, and it can catalyze the sulfate conjugation of many hormones, neurotransmitters, drugs, and xenobiotic compounds.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty