Would you like to log in again?
Human Recombinant Sulfotransferase 1A1 (from E. coli)
Supplier: ProSci
92-136
75790-866EA 946.47 USD InStock
75790-866
Human Recombinant Sulfotransferase 1A1 (from E. coli)
Enzymes
Sulfotransferase 1A1 (SULT1A1) is a cytosolic sulfotransferases that is expressed in the liver, lung, adrenal, brain, platelets, and skin
This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: N-6 His tag
SULT1A1 is a phenol sulfotransferases with thermostable enzyme activity. SULT1A1 utilizes 3'-phospho-5'-adenylyl sulfate (PAPS) as sulfonate donor to catalyze the sulfate conjugation of catecholamines, phenolic drugs and neurotransmitters. It is responsible for the sulfonation and activation of minoxidil. SULT1A1 mediates the metabolic activation of carcinogenic N-hydroxyarylamines to DNA binding products and could so participate as modulating factor of cancer risk.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty