Would you like to log in again?
Human Recombinant Peptidase D (from E. coli)
Supplier: ProSci
92-214
75791-022EA 946.47 USD InStock
75791-022
Human Recombinant Peptidase D (from E. coli)
Enzymes
PEPD belongs to the peptidase M24B family of Eukaryotic-type prolidase subfamily
This recombinant protein can be used for biological assays. For research use only.
PEPD is a cytosolic dipeptidase that hydrolyzes dipeptides with proline or hydroxyproline at the carboxy terminus. It is important in collagen metabolism because of the high levels of imino acids. Defects in PEPD are a cause of prolidase deficiency which is an autosomal recessive disorder associated with iminodipeptiduria.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty