Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Human Recombinant Magnesium-dependent phosphatase 1 (from Cells)
Supplier: ProSci
92-327
75791-240EA 946.47 USD InStock
75791-240
Human Recombinant Magnesium-dependent phosphatase 1 (from Cells)
Enzymes
MDP1, which is short for nesium-dependent phosphatase 1, is a 176 aa
This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
protein. It has 3 forms by different alternative splicing. This protein belongs to the HAD-like hydrolase superfamily, and usually takes its function with Magnesium. It can be inhibited by vanadate and zinc, and slightly by calcium. MDP1 is a Magnesium-dependent phosphatase which may act as a tyrosine phosphatase.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty