Would you like to log in again?
Human Recombinant D-tyrosyl-tRNA(Tyr) deacylase 1 (from Cells)
Supplier: ProSci
92-377
75791-340EA 946.47 USD InStock
75791-340
Human Recombinant D-tyrosyl-tRNA(Tyr) deacylase 1 (from Cells)
Enzymes
D-tyrosyl-tRNA(Tyr) deacylase 1(DTD1) belongs to the DTD family, and expressed in many adult and fetal tissues such as testis, ovary, spleen in adult and fetal brain
This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
It is a nucleus and cytoplasm located protein, and is preferentially phosphorylated in cells arrested early in S phase. DTD1 is an ATPase involved in DNA replication, it may facilitate loading of CDC45 onto pre-replication complexes. The protein may hydrolyze D-tyrosyl-tRNA(Tyr) into D-tyrosine and free tRNA(Tyr), a possible defense mechanism against a harmful effect of D-tyrosine.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty