Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Human Recombinant Thiamin pyrophosphokinase 1 (from Cells)
Supplier: ProSci
92-378
75791-342EA 629.48 USD InStock
75791-342
Human Recombinant Thiamin pyrophosphokinase 1 (from Cells)
Enzymes
Thiamin pyrophosphokinase 1 (TPK1), belongs to the thiamine pyrophosphokinase family
This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
TPK1 exists as a homodimer, and is detected in heart, kidney, testis, small intestine and peripheral blood leukocytes, and at very low levels in a variety of tissues. TPK1 can catalyze the phosphorylation of thiamine to thiamine pyrophosphate. It can also catalyze the phosphorylation of pyrithiamine to pyrithiamine pyrophosphate.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty