Would you like to log in again?
Human Recombinant Selenophosphate synthase 1 (from Cells)
Supplier: ProSci
92-396
75791-362EA 946.47 USD InStock
75791-362
Human Recombinant Selenophosphate synthase 1 (from Cells)
Enzymes
Selenophosphate synthetase 1 (SEPHS1) belongs to the selenophosphate synthase 1 family, Class II subfamily
This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
It has four different isoforms by alternative splicing. Isoform 1 and isoform 2 are gradually expressed during the cell cycle until G2/M phase and then decreased, which Isoform 3 is gradually expressed during the cell cycle until S phase and then decreased. SEPHS1 can be activated by phosphate ions and by potassium ions. It can synthesize synthesizes selenophosphate from selenide and ATP. Selenophosphate is the selenium donor used to synthesize selenocysteine, which is co-translationally incorporated into selenoproteins at in-frame UGA codons.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty