Would you like to log in again?
Mouse Recombinant Carboxypeptidase A4
Supplier: ProSci
92-415
75791-400EA 946.47 USD InStock
75791-400
Mouse Recombinant Carboxypeptidase A4
Enzymes
Carboxypeptidase A4 (CPA4) is a member of the peptidase M14 family
This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
CPA4 is metalloprotease that could be involved in the histone hyperacetylation pathway. CPA4 binds one zinc ion per subunit and could catalyze to release of a C-terminal amino acid, with preference for -Phe, -Leu, -Ile, -Met, -Tyr and -Val.They have distinct expression patterns and different specificities for example, preferentially cleaving aromatic (carboxypeptidase As) or basic (carboxypeptidase Bs) residues. Several, such as carboxypeptidase Xs, have lost their catalytic activity. Carboxypeptidases play important roles in digestion of food, processing of bioactive peptides and blood coagulation.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty