Would you like to log in again?
Mouse Recombinant Carboxylesterase 3
Supplier: ProSci
92-535
75788-718EA 912.46 USD InStock
75788-718
Mouse Recombinant Carboxylesterase 3
Enzymes
Mouse Carboxylesterases 3 (CES3) is a member of five families of mammalian carboxylesterases that plays a role in catalyzing hydrolytic and transesterification reactions with xenobiotics, anticancer pro-drugs and narcotics, and detoxifying organophosphates and insecticides
This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Mammalian carboxylesterases are enzymes with broad substrate specificities ranging from small molecule esters to longchain fatty acid esters. It is shown that CESs has key roles in the metabolism of a wide variety of clinical drugs, illicit narcotics and chemical nerve agents. CES3 is broadly expressed in liver, colon and brain.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty