Would you like to log in again?
E. coli Recombinant Tryptophan synthase (from E. coli)
Supplier: ProSci
92-667
75791-896EA 744.04 USD InStock
75791-896
E. coli Recombinant Tryptophan synthase (from E. coli)
Enzymes
Tryptophan synthase is a multienzyme alpha2 beta 2 complex composed of two protein subunit
This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: N-6 His tag
Tryptophan synthase catalyzes the last two steps in the synthesis of L-tryptophan (L-Trp). The alpha-subunit catalyzes cleavage of 3-indole-d-glycerol 3′-phosphate (IGP) to give indole and D-glyceraldehyde 3′-phosphate (G3P). Indole is then transferred through a 25-Å hydrophobic tunnel to the beta -subunit. The beta 2 subunit contains pyridoxal 5'-phosphate and catalyzes several pyridoxal 5'-phosphate-dependent reactions, including/3-elimination reactions 6 and a thiol-dependent transamination reaction. This enzyme is commonly found in Eubacteria, Archaebacteria, Protista, Fungi, and Plantae, but is absent from Animalia. As humans do not have tryptophan synthase, this enzyme has been explored as a potential drug target.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty