Would you like to log in again?
Recombinant 3-Hydroxykynurenine Transaminase Anopheles gambiae (from E. coli)
Supplier: ProSci
90-358
75794-310EA 827.54 USD InStock
75794-310
Recombinant 3-Hydroxykynurenine Transaminase Anopheles gambiae (from E. coli)
Enzymes
The 3-hydroxykynurenine transaminase (3-HKT) gene plays a vital role in the development of malaria parasites by participating in the synthesis of xanthurenic acid, which is involved in the exflagellation of microgametocytes in the midgut of malaria vector species
This recombinant proteins is for research use only.
Fusion-Tag: Tag Free
The 3-HKT enzyme is involved in the tryptophan metabolism of Anophelines. The gene had been studied in the important global malaria vector, Anopheles gambiae.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty