Would you like to log in again?
Precision High-Performance Incubators, Thermo Scientific
Supplier: Thermo Scientific
PR205075M PR205075G
97018-284EA 10680.12 USD InStock
97018-284 97018-300
Precision High-Performance Incubators, Thermo Scientific
Incubators Standard Incubators
These mechanical or gravity convection incubators with advanced microprocessor controls are ideal for applications requiring excellent temperature distribution or gentle sample heating between 5°C above ambient to 75°C.
  • Uniform heating, precise temperature control and fast drying
  • Sophisticated microprocessor PID controls with easy-to-view digital LED readout
  • Fixed setpoints on control panel eliminate need for tuning
  • Offset feature enables easy calibration
  • Built-in safety back-up maintains control at 3 °C above setpoint if primary heater control fails
  • Visual alarm indicates when temperature exceeds 3 °C setpoint
  • Double-door design with durable, enamel-coated steel exterior
Blower circulates heated air in a horizontal airflow pattern for efficient heat distribution with tight temperature tolerances of up to ±0.5 °C
Inner glass door permits viewing of samples without disturbing chamber environment in 317 L unit. Silicone gasket on outer door and 7.6 cm (3") thick fiberglass insulation prevents heat loss for high temperature uniformity. Circuit breaker protects against power surges and internal electrical outlet allows operation of a shaker, stirrer or other lab apparatus. The low-watt density heater elements are designed for long life.
Temperature is displayed on a large, three-character screen; can be easily set from 5 °C above ambient to up to 75 °C (depending on model) in 0.1 °C increments using touch-sensitive arrow keys. Gravity convection offers gentle drying with heat-generated convection to move air vertically through the chamber to heat samples.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty