Would you like to log in again?
Etest® Automated Applicator Simplex C76, Biomerieux
Supplier: bioMérieux
559802
10500-470EA 9674.8 USD InStock
10500-470
Etest® Automated Applicator Simplex C76, Biomerieux
Microbiology Test Systems Microorganism Tests
Simplex C76 automates the placement of 1 to 6 different Etest strips to simplify the setup of MIC panels.
  • Ease of use
  • Reduces operator fatigue
Allows easier application of samples to testing equipment.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty