Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Indicating Oxygen Trap, Restek

Supplier: Restek
22011 22010
10058-710EA 338.62 USD InStock
10058-710 10058-708
Indicating Oxygen Trap, Restek
Filters Gas Filters
Oxygen traps are effective at preventing oxygen and water from entering GC systems.

  • Indicator changes from light green to grey as oxygen is trapped
  • Heavy-walled glass body, protected by polycarbonate sleeve, prevents oxygen and water infusion
  • Prepurged for fast stabilization
  • 100 psi (689 kPa) maximum operating pressure
  • Reduces oxygen to 0.1 ppm
  • 10 µm frits at inlet and outlet
  • Optimal flow rate: < 150 ml/min

Delivery information: Items cannot ship FedEx ground.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty