Would you like to log in again?
Vu-Union Vespel®/Graphite Ferrules, Restek
Supplier: Restek
20421 20420 20423 20422
RK20420PK 91.48 USD InStock
RK20420 RK20422 RK20421 RK20423
Vu-Union Vespel®/Graphite Ferrules, Restek
Fittings and Connectors
These ferrules have a maximum operating temperature of 400°C.
  • Use with vacuum Vu-Union connectors
  • 60% Vespel®/40% graphite
  • 400°C maximum operating temperature
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty