Would you like to log in again?
FID Flow Measuring Adapter for Agilent 5890/6850/6890/7890 GCs, Restek
Supplier: Restek
21000
10058-588EA 101.24 USD InStock
10058-588
FID Flow Measuring Adapter for Agilent 5890/6850/6890/7890 GCs, Restek
Chromatography Supplies Chromatography Spare Parts
FID flow measuring adapters are for use with Agilent 5890/6850/6890/7890 GCs.
  • Meets or exceeds original manufacturer's performance
  • Makes setting flows easy
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty