Would you like to log in again?
Siltek® Treated Inlet Support Springs for Bruker/Varian 1075/1077 Split Injectors, Restek
Supplier: Restek
21690
RK21690PK 28.23 USD InStock
RK21690
Siltek® Treated Inlet Support Springs for Bruker/Varian 1075/1077 Split Injectors, Restek
Springs
Inlet support springs are Siltek® treated to eliminate sample adsorption.
For use with Bruker/Varian 1075/1077 split injectors.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty