Would you like to log in again?
High-Capacity Split Vent Trap, Restek
Supplier: Restek
20698 20699
RK20699PK 293.92 USD InStock
RK20699 RK20698
High-Capacity Split Vent Trap, Restek
Filters Gas Filters
High-capacity split vent traps reduce the release of hazardous materials from the capillary split vent into the lab.
Traps are sold individually or in packs of five depending on model.
Delivery information: Includes connecting lines and mounting kit
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty