Would you like to log in again?
Ridgid® Tubing Cutter, Restek
Supplier: Restek
23011
10058-838EA 91.33 USD InStock
10058-838
Ridgid® Tubing Cutter, Restek
Cutters
Excellent for cutting 1/8 or 1/4" metal tubing.
  • Compact size is ideal for tight spaces
  • Replaceable cutting wheel
Caution: Not for use with stainless steel tubing.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty