Would you like to log in again?
Moisture Control By-Pass Lines for Tekmar Instruments, Restek
Supplier: Restek
21109 21035
10855-890EA 37.35 USD InStock
10855-890 10855-876
Moisture Control By-Pass Lines for Tekmar Instruments, Restek
Tubing Transfer Tubing
Moisture control bypass lines increase the response delivered for ketones, alcohols, and acetates.
  • Silcosteel®-deactivated tubing for increased inertness
  • Suitable for U.S. EPA methods 8260, 524.2, and OLM4.1
  • Easily attaches in minutes
For use with Tekmar 3100 concentrators.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty