Would you like to log in again?
SSI TC-20 Tube Cutting Machine, Restek
Supplier: Restek
23029
10852-832EA 3490.84 USD InStock
10852-832
SSI TC-20 Tube Cutting Machine, Restek
Cutters
The SSI TC-20 cuts 1/16", 1/8", or 1/4" tubing with inside diameter as small as 0.008".
  • Electrically operated benchtop model
  • Voltage selectable 110 – 120/220 – 240 volts, 50 – 60 Hz
Handy 1/16" dressing tool on the swing arm removes burrs and reams tubing.
Delivery information: Unit shipped set for 110 – 120 operating voltage. Switch to 220 – 240 volts by using alternate fuse and power cord (included). Also replacement cutting wheels available.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty