Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Versa Vials™, Polypropylene, Restek

Supplier: Restek

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Sort all by:

22705 22706
10854-140PK 29.53 USD InStock
10854-140 10854-360
Versa Vials™, Polypropylene, Restek
Vials
Versa Vials™ feature a 12 mm cap size and durable polypropylene fabrication, with clear sides for easy observation and identification of contents.

  • Clear color
  • Polypropylene fabrication
  • 12 mm orifice

Polypropylene Versa Vials™ have a volume capacity of 1.5 ml.

Order Now
 

ORDER

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty