Would you like to log in again?
EZ Twist Top® Split/Splitless Injection Port for Agilent GCs with Large Canister-Type Filter, Restek
Supplier: Restek
22735 22736
10856-072EA 1107.9 USD InStock
10856-072 10856-074
EZ Twist Top® Split/Splitless Injection Port for Agilent GCs with Large Canister-Type Filter, Restek
Chromatography Supplies Chromatography Spare Parts
Gas lines are attached to the shell weldment bottom to help eliminate broken gas lines.
  • Change inlet liners faster and easier
  • Gas lines don't interfere with routine maintenance
  • Weldment stays secure in the tool for reattachment
Weldment removal tool allows for quick removal of the hot weldment without fingers ever touching hot surfaces. For use with large canister-type filters.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty