Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Male Connector, Parker, Restek
Supplier: Restek
22891 21945 21842 22890 21843 21942 21552 22889 21553 22887 21551 21845 21941 21554 21943 21555 21844 21944
10855-596EA 69.17 USD InStock
10855-596 10855-244 10855-594 10855-242 10855-592 RK21842 RK21843 RK21942 10854-510 10854-512 10854-720 10854-714 10854-504 10854-716 10854-506 10854-718 10854-508 10855-566
Male Connector, Parker, Restek
Fittings and Connectors
Parker's fittings are ideal for installing new equipment, modifying existing instrumentation, or replacing worn connections.
  • Fittings are available in brass or stainless steel
  • Can be treated with Siltek®/Sulfinert or Silcosteel®-CR
Compatible with a variety of connection sizes.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty