Would you like to log in again?
Hi-Duty Tubing Cutter, Restek
Supplier: Restek
22356
10058-754EA 114.83 USD InStock
10058-754
Hi-Duty Tubing Cutter, Restek
Cutters
Specifically designed to cut 1/8 to 11/8" (4 to 28 mm) OD tubing.
  • Easily cuts stainless steel, Monel alloy, and other hard-temper materials
  • Unique design makes clean, right-angle cuts and eliminates spiraling
  • Integral reamer folds away for safety and convenience
Ordering information: Includes replacement cutter wheels.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty