Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Magnetic Seals w/Septa, 20 mm, preassembled, Restek

Supplier: Restek
22443 22834 22441 22444 22832 22442 22831 22836 22835 22833
10854-116PK 878.88 USD InStock
10854-116 10854-118 10058-798 10058-778 10058-800 10058-802 10854-154 10854-120 10854-156 10854-158
Magnetic Seals w/Septa, 20 mm, preassembled, Restek
Closures Chromatography Vial Closures
Magnetic seals with variable mouth sizes are compatible with a range of vials with 20-millimeter orifices.

  • Aluminum/steel or steel fabrication
  • Compatible with crimp-top vials
  • 8 mm opening

BiMetal magnetical seals are easier to crimp and decap than standard 20 mm magnetic seals.

Ordering information: 5 mm hole is compatible with the following systems: Carlo Erba HS500/HS800, CTC 500, Fisons HS500/HS800, and Bruker/Varian/Chrompack 9020/25. 8 mm hole is compatible with the Combi Pal.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty