Would you like to log in again?
Replacement Ring Seal for MSD Conversion Fitting, Restek
Supplier: Restek
21313
RK21313PK 20.46 USD InStock
RK21313
Replacement Ring Seal for MSD Conversion Fitting, Restek
Seals Sealing Rings
Flat, soft ring seal deforms and butt-seals against MSD interface.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty