Would you like to log in again?
AMBITEX® Vinyl Powder-Free Exam Gloves, V200 Series, Cardinal Health®
Supplier: Cardinal
AMBITEX®
VMD200 VXL200 VLG200
10861-016BX 13.35 USD InStock
10861-016 10860-948 75779-666
AMBITEX® Vinyl Powder-Free Exam Gloves, V200 Series, Cardinal Health®
Gloves
Powder-free.
  • Ideal for most tasks involving minimal risk of biological contamination
  • Cost-effective when frequent glove changes required
  • Smooth surface
  • Not made with natural rubber latex
Certifications: Exceeds ASTM and FDA examination glove standards. Manufactured in an ISO 9001 certified facility. Complies with 21 CFR parts 170-199 for use in contact with food.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty