Would you like to log in again?
SP Bel-Art H-B Instrument DURAC Plus ASTM Like Liquid-In-Glass Thermometers, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a part of SP
B60750-6600 B60750-7000
75795-852CS 723.49 USD InStock
75795-852 75795-854
SP Bel-Art H-B Instrument DURAC Plus ASTM Like Liquid-In-Glass Thermometers, Bel-Art Products, a part of SP
Thermometers Analog Thermometers
Mercury Free, these ASTM Like thermometers are safer to use than their mercury counterparts.
  • Mercury-free
  • Lead-free white back glass
  • Blue organic liquid fill
Ideal for use in quality control laboratories or when mercury filled thermometers cannot be used, but ASTM standards are still needed
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty