Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Procter & Gamble Joy® Dishwashing Liquid
Supplier: T. Frank McCall's
Joy®
68003JOY38OZ
42000-151CS 205.79 USD InStock
42000-151
Procter & Gamble Joy® Dishwashing Liquid
Cleaning Agents Detergents and Surface Cleansers
Powerful and concentrated surfactant to clean a wide variety of soils.
  • Designed to be mild
  • High surfactant level to give a rich layer of suds and high suds mileage
Forms surfactant micelles that stay suspended in wash water to provide microreservoirs of reserve cleaning power. Use 29.5 to 88.72 ml (1 to 3 fl.oz.) per 37.85 L (10 gal.) sink.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty