Would you like to log in again?
PromoFectin-HUVEC, PromoKine
Supplier: PromoKine
CT-2000-HUV-50 CT-2000-HUV-10
10181-144EA 206.96 USD InStock
10181-144 10181-146
PromoFectin-HUVEC, PromoKine
Transfection Reagents
PromoFectin-HUVEC is ideally suited for transfection of endothelial cells.
  • Less cell stress
  • Better cell viability
  • Up to 50% transfection rates could be achieved with primary endothelial cells
PromoFectin-HUVEC is a variant of PromoKine's standard transfection reagent PromoFectin. Using PromoFectin-HUVEC up to 50% transfection rates could be achieved with primary endothelial cells such as HUVEC and HDMEC and it may be better suited for this kind of fragile cell type than the broad-range PromoFectin. PromoFectin-HUVEC is ideally suited for transfection of endothelial cells (also vascular endothelial cells of other origin) - yielding almost the same transfection rates than with our MATra technology but being more gentle to these very sensitive cells. This results in less cell stress and better cell viability (i.e. vitality). While PromoFectin can typically remain on most cells without media changes and washes, we anyway recommend medium change when working with very sensitive cells.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty