Would you like to log in again?
Gifu Anaerobic Broth, Himedia Laboratories
Supplier: HiMedia Laboratories
M1801-5KG M1801-5KG
89224-698EA 1673.86 USD InStock
CA89224-698 89224-698
Gifu Anaerobic Broth, Himedia Laboratories
Media for Microbiology Prepared Media for Microbiology
Gifu Anaerobic Medium (GAM Broth) is a liquid medium for anaerobic bacteria.
Gifu anaerobic medium is recommended as a general culture medium for cultivation and isolation of anaerobic bacteria and to test their susceptibility to antibiotics other than sulpha drugs. This medium is also suitable for blood culture.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty