Would you like to log in again?
Optima Bench Cart, PerkinElmer
Supplier: Perkin Elmer
N0771725
10138-120EA 9215.52 USD InStock
10138-120
Optima Bench Cart, PerkinElmer
Carts
Optima bench for optical emission spectrometer.
  • Heavy-duty design
  • Locking casters
  • Cost-effective
  • Additional shelf for organizing consumables
This basic bench is a cost-effective option for storing Optima instruments. With an adjustable shelf designed for an autosampler, the autosampler can be configured to a position that is best suited. This bench also comes with a half storage shelf for organizing consumables and supplies.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty