Would you like to log in again?
Live/Dead Cell Staining Kit, MilliporeSigma
Supplier: MilliporeSigma
QIA76-100TEST
EMQIA76100TESTEA 865.47 USD InStock
EMQIA76100TEST
Live/Dead Cell Staining Kit, MilliporeSigma
Assays Cellular Assays
The Live/Dead Cell Staining Kit utilizes two fluorescent dyes, Cyto-dye to stain live cells and propidium iodide to stain dead cells.
  • Utilizes two fluorescent dyes
  • Band-pass filter which detects FITC and rhodamine
Stained live and dead cells can be visualized by fluorescence microscopy using a band-pass filter which detects FITC and rhodamine. Viable cells will stain only with the Cyto-dye, fluorescing green, whereas the dead cells will stain with both Cyto-dye (green) and propidium iodide (red), resulting in a yellow fluorescence.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty