Would you like to log in again?
Ultrafree-MC GV Centrifugal Filter, MilliporeSigma
Supplier: MilliporeSigma
UFC30GV0S
102970-584PK 355.38 USD InStock
102970-584
Ultrafree-MC GV Centrifugal Filter, MilliporeSigma
Filters Centrifugal Filters
Ultrafree® centrifugal devices are single-use, disposable filters used for removing particulates from aqueous biological solutions.
  • Five different pore sizes from 0.1 to 5.0 µm
  • Fast filtration and highly reproducible performance
  • Use in fixed-angle rotors for 1.5 ml tubes
These filters are used for the removal of particles and precipitates from aqueous and some solvent-based samples. Ideal for use with protein and nucleic acid solutions.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty