Would you like to log in again?
TGF-beta2 Human ELISA, BioVendor
Supplier: BioVendor
RAF125R
102552-586EA 959.14 USD InStock
102552-586
TGF-beta2 Human ELISA, BioVendor
Assays ELISAs
TGF-β2 is a potent cytokine which has been shown to modulate embryonic development.
  • Calibration Range: 31.3-1000 pg/mL
  • Limit of Detection: 6.6 pg/mL
  • Cell culture supernatant, Serum: 100 µL (1:100 prediluted)
TGF-beta belongs to a family of dimeric 25 kDa polypeptides that are ubiquitously distributed in tissues and synthesized by many different cells. TGF-β2 is a potent cytokine which has been shown to modulate embryonic development, bone formation, mammary development, wound healing, hematopoiesis, cell cycle progression and the production of the extracellular matrix.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty