Would you like to log in again?
Millikan Apparatus
Supplier: KLINGER EDUCATION PROD CORP SE
P6121
470220-110KT 7071.09 USD InStock
470220-110
Millikan Apparatus
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
Measure suspension voltage and falling speed.
  • Includes all major components
  • Repeatable and precise
In the experiment the electric field is calculated from the voltage U at the plate capacitor at which the observed oil droplet just begins to hover. The constant falling velocity v1 of the droplet when the electric field is switched off is subsequently measured to determine the radius. Repeating the experiment allows student to determine the mass of an electron charge.
Millikan apparatus
Millikan supply unit
Electronic time clock P
Connecting lead 19 A, 100 cm, red/blue, pair (3)
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty