Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Zeeman Effect

Supplier: KLINGER EDUCATION PROD CORP SE
P6274B
470220-120KT 25097.09 USD InStock
470220-120
Zeeman Effect
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
Measuring the Zeeman splitting of red cadmium.

  • Includes all major components
  • Observe the Cadmium line

In this experiment the Zeeman splitting of the red cadmium line is measured as a function of the magnetic field B. The energy interval of the triplet components is used to calculate the specific electron charge.

Required but not included: PC with Windows 2000/XP/Vista/7/8/10 (x86 or x64).

Spectral lamp Cd, with holding plate
Universal choke
U-core with yoke
Coil 480 turns, 10 A (2)
Pole pieces with large bore, pair
High current power supply
Fabry-Perot-Etalon
Lens in frame, f = +150 mm (2)
Holder for interference filters
Interference filter, 644 nm
VideoCom USB
Optical bench, standard cross section 1 m
Rider base with threads
Optics rider 60/50 (5)
Mobile-CASSY
Combi B sensor S
Extension cable, 15-pole
Stand base V-shape, small
Stand rod 47 cm, 12 mm Ø
Leybold multiclamp
Connecting lead 32 A, 100 cm, black (3)

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty