Would you like to log in again?
Mechanical Effect Demonstration
PH1212
470159-560EA 42.65 USD InStock
470159-560
Mechanical Effect Demonstration
Educational Materials Physics Educational Materials Mechanics Learning Activities
Visualize modern physics by watching an electron beam creating motion on a paddle wheel.
  • Demonstration Crooke's tube
  • Clearly visible beam
The magnetic effect demonstration shows that cathode rays have kinetic energy, by making the blade wheel rotate! Cathode Rays exert a pushing effect on the object that it falls on, which is shown by the rotation in the demonstration.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty