Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Distillation Receivers, Graduated, Chemglass
Supplier: Chemglass
CG-1272-01
80067-856EA 1107.5 USD InStock
80067-856
Distillation Receivers, Graduated, Chemglass
Distillation Components Distillation Receivers
Graduated distillation receiver with a 4 mm three way stopcock to permit solvent removal through the 10 mm outer diameter tube or return to distillation flask
Upper sampling port has a 2 mm PTFE stopcock. The top outer and lower inner joints are the same size. Solvent recovery with 24/40 joint size.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty