Would you like to log in again?
Steris Spordex Culture Media
Supplier: Steris
NA114 NA114
48350-012PK 505.12 USD InStock
CA48350-012 48350-012
Steris Spordex Culture Media
Media for Microbiology Prepared Media for Microbiology
This media used with Steris Spordex and Spordi biological indicator strips consists of a modified soybean casein digest broth that contains a pH indicator.
  • Consists of a modified soybean casein digest broth that contains a pH indicator
  • Easy-to-read color change in the medium
This media used with Steris Spordex and Spordi biological indicator strips consists of a modified soybean casein digest broth that contains a pH indicator. An easy-to-read color change in the medium to yellow is indicative of microbial growth.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty