Would you like to log in again?
HQ440d Benchtop Meter Package with PHC108 pH Ultra Electrode, Hach
Supplier: Hach
8506250
10146-108EA 6259.7 USD InStock
10146-108
HQ440d Benchtop Meter Package with PHC108 pH Ultra Electrode, Hach
Electrochemical Meters pH and Multi-Parameter Meters
Designed for water quality experts, the Hach HQ440d benchtop meter is an advanced laboratory meter that takes the guesswork out of measurements.
  • An intuitive user interface for simple operation and accurate results
  • With IntelliCAL™ smart probes that store all calibrations in the probe
  • Enhance productivity with an ultra-bright screen and large font size for easy to read results
HQd meters connect with smart probes that automatically recognize the testing parameter, calibration history, and method settings to minimize errors and setup time. The advanced benchtop meter is designed to withstand years of continuous lab use. HQd meters feature a simple user interface that does not require manuals or training to operate. The high-flowing reference and open junction of the pH Ultra electrode included in this package delivers fast stablilzation time even in the most difficult sample types and low ionic strength waters.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty