Would you like to log in again?
Adapter, pH Probes, Chemglass
Supplier: Chemglass
CG-1872-A-10
16002-804EA 308.8 USD InStock
16002-804
Adapter, pH Probes, Chemglass
Adapters Adapters, Joints with Tube Connectors
Adapter permits pH electrode to be inserted either directly into smaller reactors or down through a baffle.
  • Compression adapter union
  • For use with probes with open bottom baffles
  • For 10L and larger reactors
Allows CG-1872 series pH electrode (12mm OD) to be inserted either directly into smaller reactors or down through a CG-1872-B baffle.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty