Would you like to log in again?
Wide Mouth Centrifuge Bottle, Chemglass
Supplier: Chemglass
CLS-1156-0400 CLS-1156-0180 CLS-1156-1000 CLS-1156-0500
10034-164CS 126.59 USD InStock
10034-164 10034-166 10034-162 10034-168
Wide Mouth Centrifuge Bottle, Chemglass
Bottles Centrifuge Bottles
These wide-mouth, polycarbonate bottles are mainly used for ultracentrifugation.
  • Bottles withstand maximum speeds without leakage and are ideal for all GS3 rotors
  • Used where relatively small amounts of solids are involved
  • Tight-fitting, polypropylene screw caps reduce the possibility of aerosol escape
  • Bottles and caps are reusable and sterilizable by autoclaving
  • Supplied nonsterile with caps attached
They are also used for culture microbes, plants, algae, and organisms requiring large surface area to volume ratios.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty